Ethyl myristate
Supplier: Cayman Chemical
Synonyms:
Tetradecanoic acid ethyl ester, Tetradecylic acid ethyl ester, Ethyl tetradecanoate, Myristic acid ethyl ester
10008197-100
10008197-500
10008197-1
CAYM10008197-100EA
33
EUR
CAYM10008197-100
CAYM10008197-500
CAYM10008197-1
Ethyl myristate
Ethyl myristate
Formula:
C₁₆H₃₂O₂ MW: 256.43 g/mol Density: 0.86 g/cm³ (20 °C) |
MDL Number:
MFCD00008984 CAS Number: 124-06-1 EINECS: 204-675-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...