Acetic acid ≥99%

Supplier: Roth Carl
Danger

7332.1 7332.2 7332.3
ROTH7332.1EA 0 EUR
ROTH7332.1 ROTH7332.2 ROTH7332.3
Acetic acid ≥99%
Acetic acid
Formula: H₃CCOOH
Storage Temperature: Ambient
MDL Number: MFCD00036152
CAS Number: 64-19-7
EINECS: 200-580-7
UN: 2789
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ecdb21d8177dd197c843d4b5187410fee102541&vid=6c26d1716b942e60cb8b3ec0f91e6623fed5b3db&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR