Lithium octanoate ≥99%

Supplier: Thermo Scientific
Warning

39371.14 39371.22
39371.14EA 20.6 EUR
39371.14 39371.22
Lithium octanoate ≥99%
Lithium octanoate

Purity: 99% CAS No.: 16577-52-9 Molecular Formula: LiO2C(CH2)6CH3 Formula Weight: 150.14 Physical Form: Powder

Formula: C₈H₁₅LiO₂
MW: 150,15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00210629
CAS Number: 16577-52-9
EINECS: 240-637-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR