Magnesium stearate
Supplier: Thermo Scientific
Synonyms:
Stearic acid Magnesium(II) salt, Magnesium(II) Stearate, Stearic acid magnesium salt
A16244.0B
A16244.30
A16244.0BEA
86.2
EUR
A16244.0B
A16244.30
Magnesium stearate
Magnesium stearate
Formula:
C₃₆H₇₀MgO₄ MW: 591,25 g/mol |
MDL Number:
MFCD00036391 CAS Number: 557-04-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...