Silver(I) acetate 99%, pure
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
Silver acetate, Acetic acid silver (I) salt
202390250
202391000
202395000
ACRO202390250EA
250
EUR
ACRO202390250
ACRO202391000
ACRO202395000
Silver(I) acetate 99%, pure
Silver (I) acetate
Formula:
C₂H₃AgO₂ MW: 166,91 g/mol Melting Pt: 2000 °C Density: 3,25 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00012446 CAS Number: 563-63-3 EINECS: 209-254-9 UN: 3077 ADR: 9,III Merck Index: 12,08648 |
Specification Test Results
Appearance | Off-white to beige to light grey Powder |
Infrared spectrum | Conforms |
Titration Argentometric | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...