Glycolic acid 99%
Supplier: Thermo Fisher Scientific
|
Danger
|
Synonyms:
2-Hydroxy acid, 2-Hydroxyacetic acid
154510250
154511000
154515000
154510025
ACRO154510250EA
47.8
EUR
ACRO154510250
ACRO154511000
ACRO154515000
ACRO154510025
Glycolic acid 99%
Glycolic acid
Formula:
C₂H₄O₃ MW: 76.05 g/mol Boiling Pt: 100 °C (760 mmHg) Melting Pt: 72…80 °C Flash Pt: >300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004312 CAS Number: 79-14-1 EINECS: 201-180-5 UN: 3261 ADR: 8 II Merck Index: 13,04511 |
Specification Test Results
Appearance | White to off-white Adhering crystals |
Infrared spectrum | Conforms |
Melting point | 72 to 80 °C |
Water | ≤1 % |
Titration with NaOH | ≥98.5 % (On dry substance) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...