Succinic acid
Supplier: Roth Carl
|
Warning
|
Synonyms:
1,4-Butanedioic acid
ROTH8268.2EA
87.6
EUR
ROTH8268.2
ROTH8268.1
Succinic acid
Succinic acid
Formula:
HOOCCH₂CH₂COOH MW: 118,09 g/mol Boiling Pt: 235 °C (1013 hPa) Melting Pt: 182 °C Density: 1,56 g/cm³ (20 °C) Flash Pt: 206 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002789 CAS Number: 110-15-6 EINECS: 203-740-4 UN: 3261 ADR: 8,III Merck Index: 13,08953 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...