Suberic acid 99%
Supplier: Thermo Fisher Scientific
|
Warning
|
132811000
132815000
132810025
ACRO132811000EA
39.7
EUR
ACRO132811000
ACRO132815000
ACRO132810025
Suberic acid 99%
Suberic acid
Formula:
C₈H₁₄O₄ Boiling Pt: 230 °C (15 mmHg) Melting Pt: 141…144 °C Density: 1,354 g/cm³ (21 °C) Flash Pt: 203 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004428 CAS Number: 505-48-6 EINECS: 208-010-9 Merck Index: 12,09031 |
Specification Test Results
Appearance | White to cream Powder |
Infrared spectrum | Conforms |
Melting point | 141 to 144 °C |
Titration with NaOH | 98.5 to 101.5 % |
Elemental Analysis | 6.22 to 7.02 % (N) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...