Disodium oxalate

Supplier: Roth Carl
Warning

Synonyms: Sodium ethanedioate, Sodium oxalate, Soerensen's buffer substances

4267.1 4267.2
ROTH4267.1EA 63.1 EUR
ROTH4267.1 ROTH4267.2
Disodium oxalate
di-Sodium oxalate
Formula: Na₂C₂O₄
MW: 134 g/mol
Melting Pt: 250…270 °C
Density: 2,27 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00012465
CAS Number: 62-76-0
EINECS: 200-550-3
Merck Index: 13,08723

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR