Diammonium oxalate

Supplier: Roth Carl
Warning

4266.1 4266.2
ROTH4266.1EA 0 EUR
ROTH4266.1 ROTH4266.2
Diammonium oxalate
di-Ammonium oxalate
Formula: C₂H₈N₂O₄
Melting Pt: 70 °C
Density: 1,5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00013308
CAS Number: 1113-38-8
EINECS: 214-202-3
UN: 2811
ADR: 6.1,III
Merck Index: 12,00571

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR