Potassium citrate monohydrate
Supplier: Roth Carl
Synonyms:
Potassium citrate tribasic monohydrate, Citric acid tripotassium salt monohydrate, Tripotassium citrate monohydrate, Tripotassium 2-hydroxypropane-1,2,3-tricarboxylate monohydrate
X888.1
X888.2
X888.3
ROTHX888.1EA
38.6
EUR
ROTHX888.1
ROTHX888.2
ROTHX888.3
Potassium citrate monohydrate
Potassium citrate monohydrate
Formula:
HOC(COOK)(CH₂COOK)₂·H₂O MW: 324.41 g/mol Melting Pt: 230 Density: 1.98 g/cm³ (20 °C) |
MDL Number:
MFCD00150442 CAS Number: 6100-05-6 EINECS: 212-755-5 Merck Index: 13,07706 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...