Thioacetic acid for synthesis, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
M-Clarity™ quality level = MQ200
8.08076.0100
8.08076.0250
8.08076.9025
8.08076.0100EA
41.3
EUR
8.08076.0100
8.08076.0250
8.08076.9025
Thioacetic acid for synthesis, Sigma-Aldrich®
Thioacetic acid
Formula:
C₂H₄OS MW: 76,12 g/mol Boiling Pt: 90 °C (1013 hPa) Melting Pt: –17 °C Density: 1,07 g/cm³ (20 °C) Flash Pt: 18 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004853 CAS Number: 507-09-5 EINECS: 208-063-8 UN: 2436 ADR: 3,II Merck Index: 12,09454 |
Specification Test Results
Solubility | 27 g/l (15 °C) (slow decomposition) |
Melting Point | -17 °C |
Molar Mass | 76.12 g/mol |
Boiling Point | 88 - 92 °C (1013 hPa) |
Vapor Pressure | 1 hPa (20 °C) |
Flash Point | 18 °C |
Refractive Index | 1.463 (20 °C, 589 nm) |
Density | 1.07 g/cm3 (20 °C) |
pH | 1.8 (100 g/l, H2O, 20 °C) Hydrolysis |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...