Thiourea ≥99% ACS
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
Thiocarbamide, Sulphourea
36609.18
36609.30
36609.A1
36609.18EA
48.2
EUR
36609.18
36609.30
36609.A1
Thiourea ≥99% ACS
Thiourea
Purity: 99% min.
CAS No.: 62-56-6
Molecular Formula: CH4N2S
Formula Weight: 76.12
Physical Form: Crystalline
Density: 1.405
Appearance: White
Odour: Odourless
Melting Point: 175-178°
Beilstein Reference: 605327
Merck Reference: 14,9367
UN No.: 3077
MDL No.: MFCD00008067
Reagent for organic synthesis
Formula:
NH₂CSNH₂ MW: 76,12 g/mol Melting Pt: 175…178 °C Density: 1,405 Storage Temperature: Ambient |
MDL Number:
MFCD00008067 CAS Number: 62-56-6 EINECS: 200-543-5 UN: 3077 ADR: 9,III Merck Index: 14,9367 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...