Trifluoroacetamide for synthesis, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
Synonyms:
2,2,2-Trifluoroacetamide
M-Clarity™ quality level = MQ200
8.14690.0010
8.14690.0050
8.14690.0010EA
33.1
EUR
8.14690.0010
8.14690.0050
Trifluoroacetamide for synthesis, Sigma-Aldrich®
Trifluoroacetamide
Formula:
C₂H₂F₃NO Boiling Pt: 162,4 °C (1013 hPa) Melting Pt: 73…75 °C Density: 1 g/cm³ (20 °C) |
MDL Number:
MFCD00008008 CAS Number: 354-38-1 EINECS: 206-559-9 |
Specification Test Results
Solubility | 460 g/l (23 °C) |
Melting Point | 73 - 75 °C |
Molar Mass | 113.03 g/mol |
Bulk Density | 960 kg/m3 |
Boiling Point | 162.5 °C |
pH | 3.6 (460 g/l, H2O, 23 °C) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...