Ethyl fluoroacetate 97%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Fluoroacetic ethyl ester

172660050 172660500
ACRO172660050EA 28.7 EUR
ACRO172660050 ACRO172660500
Ethyl fluoroacetate 97%
Ethyl fluoroacetate
Formula: C₄H₇FO₂
MW: 106.1 g/mol
Boiling Pt: 119.2 °C (753 mmHg)
Density: 1.09 g/cm³
Flash Pt: 30 °C
MDL Number: MFCD00000450
CAS Number: 459-72-3
EINECS: 207-297-8
UN: 2929

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3745 to 1.3765 (20°C, 589 nm)
GC ≥96.0 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR