Ethyl trifluoroacetate ≥99%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Trifluoroacetic ethyl ester

A11520.0B A11520.18 A11520.30
A11520.0BEA 757 EUR
A11520.0B A11520.18 A11520.30
Ethyl trifluoroacetate ≥99%
Ethyl trifluoroacetate

Purity: 99% CAS No.: 383-63-1 Molecular Formula: C4H5F3O2 Formula Weight: 142.08 Flash Point: -1°(30°F) Refractive Index: 1.3070 Odour: Sweet Melting Point: -78° Boiling Point: 60-62° Beilstein Reference: 1761411 UN No.: 2924 MDL No.: MFCD00000419

Formula: C₄H₅F₃O₂
Boiling Pt: 60…62 °C
Melting Pt: < –78 °C
Density: 1,192
Flash Pt: –1 °C (30 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000419
CAS Number: 383-63-1
EINECS: 206-851-6
UN: 2924
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR