2,2-Difluorobutyric acid ≥97%

Supplier: Thermo Scientific
Danger

Synonyms: 2,2-Difluorobutanoic acid

H25830.MD H25830.03
H25830.MDEA 97.7 EUR
H25830.MD H25830.03
2,2-Difluorobutyric acid ≥97%
2,2-Difluorobutyric acid

Purity: 97% CAS No.: 2366-62-3 Molecular Formula: C4H6F2O2 Formula Weight: 124.09 Refractive Index: 1.3645 Melting Point: 7-9° Boiling Point: 100-101°/8.5mm UN No.: 3265 MDL No.: MFCD03093765

Formula: C₄H₆F₂O₂
MW: 124,09 g/mol
Boiling Pt: 100…101 °C (8,5 mmHg)
Melting Pt: 7…9 °C
Density: 1,220
Storage Temperature: Ambient
MDL Number: MFCD03093765
CAS Number: 2366-62-3
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR