Phenyl trifluoroacetate ≥98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
Trifluoroacetic phenyl ester
B21304.06
B21304.14
B21304.06EA
28
EUR
B21304.06
B21304.14
Phenyl trifluoroacetate ≥98%
Phenyl trifluoroacetate
Purity: 98%
CAS No.: 500-73-2
Molecular Formula: C6H5OCOCF3
Formula Weight: 190.12
Flash Point: 52°(127°F)
Refractive Index: 1.4230
Boiling Point: 146-147°
UN No.: 3272
MDL No.: MFCD00039232
Formula:
C₈H₅F₃O₂ MW: 190,12 g/mol Boiling Pt: 146…147 °C Melting Pt: –8,5 °C Density: 1,276 Flash Pt: 52 °C (127 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00039232 CAS Number: 500-73-2 EINECS: 207-911-4 UN: 3272 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...