Perfluoro-tert-butanol ≥99%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
Perfluoro-t-butanol, Nonafluoro-tert-butyl alcohol, 1,1,1,3,3,3-Hexafluoro-2-trifluoromethyl-2-propanol, Perfluoro-tert-butyl alcohol
B22442.03
B22442.06
B22442.14
B22442.03EA
65.2
EUR
B22442.03
B22442.06
B22442.14
Perfluoro-tert-butanol ≥99%
Perfluoro-tert-butanol
Purity: 99%
CAS No.: 2378-02-1
Molecular Formula: C4HF9O
Formula Weight: 236.04
Flash Point: None
Refractive Index: 1.3 00
Odour: Characteristic
Melting Point: -17°
Boiling Point: 44-45°
Beilstein Reference: 1841640
UN No.: 2810
MDL No.: MFCD00042092
Formula:
C₄HF₉O MW: 236,04 g/mol Boiling Pt: 44…45 °C Melting Pt: –17 °C Density: 1,693 Storage Temperature: Ambient |
MDL Number:
MFCD00042092 CAS Number: 2378-02-1 EINECS: 219-157-3 UN: 2810 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...