N,N-Dimethylformamide diethyl acetal 95%
Supplier: Thermo Fisher Scientific
|
Danger
|
Synonyms:
1,1-Diethoxy-N,N-dimethylmethanamine, 1,1-Diethoxytrimethylamine
L00205.09
L00205.18
L00205.09EA
38.2
EUR
L00205.09
L00205.18
N,N-Dimethylformamide diethyl acetal 95%
N,N-Dimethylformamide diethyl acetal
Purity: 95%
CAS No.: 1188-33-6
Molecular Formula: (CH3)2NCH(OCH2CH3)2
Formula Weight: 147.22
Flash Point: 17°(62°F)
Refractive Index: 1.3990
Boiling Point: 134-136°
Beilstein Reference: 741889
UN No.: 1993
MDL No.: MFCD00009227
Formula:
C₇H₁₇NO₂ Boiling Pt: 134…136 °C Density: 0,859 Flash Pt: 17 °C (62 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009227 CAS Number: 1188-33-6 EINECS: 214-707-9 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...