N,N-Dimethyloctylamine for synthesis, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
M-Clarity™ quality level = MQ200
8.41863.0025
8.41863.0100
8.41863.0002
8.41863.0025EA
53.6
EUR
8.41863.0025
8.41863.0100
8.41863.0002
N,N-Dimethyloctylamine for synthesis, Sigma-Aldrich®
N,N-Dimethyloctylamine
Formula:
C₁₀H₂₃N Boiling Pt: 195 °C (1013 hPa) Melting Pt: –57 °C Density: 0,765 g/cm³ (20 °C) Flash Pt: 65 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009558 CAS Number: 7378-99-6 EINECS: 230-939-3 UN: 2922 ADR: 8 (6.1),II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...