1,4-Diisocyanatobutane 97%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
Tetramethylene diisocyanate
314380010
314380050
314380250
ACRO314380010EA
99.8
EUR
ACRO314380010
ACRO314380050
ACRO314380250
1,4-Diisocyanatobutane 97%
1,4-Diisocyanatobutane
Formula:
C₆H₈N₂O₂ MW: 140,14 g/mol Boiling Pt: 102…104 °C (14 mmHg) Melting Pt: 185…187 °C Density: 1,1 g/cm³ Flash Pt: 106 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00075559 CAS Number: 4538-37-8 UN: 2206 ADR: 6.1,III |
Specification Test Results
Appearance | Clear light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4510 to 1.4530 (20°C, 589 nm) |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...