6-Aminohexan-1-ol 94%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 6-Amino-1-hexanol

294600100 294600500
ACRO294600100EA 123 EUR
ACRO294600100 ACRO294600500
6-Aminohexan-1-ol 94%
6-Aminohexan-1-ol
Formula: C₆H₁₅NO
Boiling Pt: 227 °C (1013 hPa)
Melting Pt: 53…59 °C
Flash Pt: 133 °C
Storage Temperature: Ambient
MDL Number: MFCD00008241
CAS Number: 4048-33-3
EINECS: 223-748-1

Order Now

Specification Test Results

Appearance White to yellow to light brown Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 53 to 59 °C
GC ≥92 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR