Dibutylamine 99%
Supplier: Thermo Fisher Scientific
|
Danger
|
Synonyms:
Di-n-butylamine, N-butyl-1-propanamine
112950010
112950025
112950100
112950500
ACRO112950010EA
110
EUR
ACRO112950010
ACRO112950025
ACRO112950100
ACRO112950500
Dibutylamine 99%
Dibutylamine
Formula:
C₈H₁₉N Boiling Pt: 159 °C (760 mmHg) Melting Pt: –62 °C Density: 0,76 g/cm³ Flash Pt: 39 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009429 CAS Number: 111-92-2 EINECS: 203-921-8 UN: 2248 ADR: 8,II Merck Index: 13,03058 |
|
Specification Test Results
Appearance | Clear liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4160 to 1.4180 (20°C, 589 nm) |
Water | ≤0.1 % |
GC | ≥98.5 % |
Color scale | ≤20 APHA |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...