2-Ethylaminoethanol for synthesis, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
M-Clarity™ quality level = MQ200
8.01392.0250
8.01392.1000
8.01392.0250EA
41.9
EUR
8.01392.0250
8.01392.1000
2-Ethylaminoethanol for synthesis, Sigma-Aldrich®
2-Ethylaminoethanol
Formula:
C₄H₁₁NO Boiling Pt: 167 °C (1013 hPa) Melting Pt: –9 °C Density: 0,914 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002841 CAS Number: 110-73-6 EINECS: 203-797-5 UN: 2735 ADR: 8,II |
Specification Test Results
Solubility | (20 °C) soluble |
Melting Point | -9 °C |
Molar Mass | 89.14 g/mol |
Boiling Point | 167 °C |
Vapor Pressure |
|
Flash Point | 78 °C |
Refractive Index | 1.4402 (20 °C, 589 nm) |
Explosion Limit | 1.6 - 11.7 %(V) |
Density | 0.914 g/cm3 (20 °C) |
pH | 12.3 (100 g/l, H2O, 20 °C) |
Ignition Point | 330 °C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...