Methanesulphonic anhydride ≥98%

Supplier: Thermo Fisher Scientific
Danger

A13328.30 A13328.18 A13328.09 A13328.14
A13328.30EA 644 EUR
A13328.30 A13328.18 A13328.09 A13328.14
Methanesulphonic anhydride ≥98%
Methanesulphonic anhydride

Purity: 98% CAS No.: 7143-01-3 Molecular Formula: C2H6O5S2 Formula Weight: 174.20 Melting Point: 65-69° Boiling Point: 125°/4mm UN No.: 3261 MDL No.: MFCD00007556

Formula: C₂H₆O₅S₂
MW: 174.2 g/mol
Boiling Pt: 125 °C (4 mmHg)
Melting Pt: 65…69 °C
Storage Temperature: Ambient
MDL Number: MFCD00007556
CAS Number: 7143-01-3
EINECS: 230-442-1
UN: 3261
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR