2-Ethoxyethanol ≥99%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Ethyl glycol, Ethylene glycol monoethyl ether, Cellosolve

A16100.0F A16100.AP
A16100.0FEA 79.2 EUR
A16100.0F A16100.AP
2-Ethoxyethanol ≥99%
2-Ethoxyethanol

Purity: 99% CAS No.: 110-80-5 Molecular Formula: C4H10O2 Formula Weight: 90.12 Flash Point: 44°(111°F) Refractive Index: 1.4085 Odour: Ether-like Melting Point: -90° Boiling Point: 134-135° Beilstein Reference: 1098271 Merck Reference: 14,3750 UN No.: 1171 MDL No.: MFCD00002869

Formula: C₂H₅OCH₂CH₂OH
Boiling Pt: 134…135 °C
Melting Pt: –90 °C
Density: 0,931
Flash Pt: 44 °C (111 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002869
CAS Number: 110-80-5
EINECS: 203-804-1
UN: 1171
ADR: 3,III
REACH: 01-2119560582-38
Merck Index: 14,3750
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=461a4daf8709ebd6b4516d73b4b32eee33cd1bca&vid=625adefb8d1e2126363931c57a51d3e1df419926&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR