3-Methoxy-1-butanol 99%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
3-Methoxybutan-1-ol
125790010
125791000
ACRO125790010EA
76.8
EUR
ACRO125790010
ACRO125791000
3-Methoxy-1-butanol 99%
3-Methoxy-1-butanol
Formula:
C₅H₁₂O₂ MW: 104,15 g/mol Boiling Pt: 161 °C (1013 hPa) Melting Pt: –85 °C Density: 0,92 g/cm³ Flash Pt: 46 °C |
MDL Number:
MFCD00002931 CAS Number: 2517-43-3 EINECS: 219-741-8 UN: 1987 ADR: 3,III |
Specification Test Results
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4150 to 1.4165 (20°C, 589 nm) |
Water | ≤0.2 % |
GC | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...