1,12-Dodecanediol 98%

Supplier: Thermo Scientific

Synonyms: 1,12-Dihydroxy dodecane

117610250 117611000 117610010
ACRO117610250EA 30.3 EUR
ACRO117610250 ACRO117611000 ACRO117610010
1,12-Dodecanediol 98%
1,12-Dodecanediol
Formula: C₁₂H₂₆O₂
MW: 202,34 g/mol
Boiling Pt: 324 °C (760 mmHg)
Melting Pt: 79…81 °C
Density: 1,08 g/cm³ (20 °C)
Flash Pt: 176 °C
Storage Temperature: Ambient
MDL Number: MFCD00004755
CAS Number: 5675-51-4
EINECS: 227-133-9

Order Now

Specification Test Results

Appearance White Crystals or flakes
Infrared spectrum Conforms
Melting point 79 to 81 °C
GC ≥97.5 %
Residue after reduction ≤0.02 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR