Ethyl methanesulphonate ≥99%
Supplier: Thermo Fisher Scientific
|
Danger
|
Purity: 99%
CAS No.: 62-50-0
Molecular Formula: CH3SO3CH2CH3
Formula Weight: 124.16
Flash Point: 100°(212°F)
Physical Form: Liquid
Density: 1.170
Refractive Index: 1.4180
Sensitivity: Moisture Sensitive
Appearance: Colourless
Odour: Fruit-like
Boiling Point: 85-86°/10mm
Beilstein Reference: 773969
Merck Reference: 14,3827
UN No.: 2810
MDL No.: MFCD00007559
Formula:
C₃H₈O₃S Boiling Pt: 85…86 °C (10 mmHg) Density: 1,170 Flash Pt: 100 °C (212 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007559 CAS Number: 62-50-0 EINECS: 200-536-7 UN: 2810 ADR: 6.1,II Merck Index: 14,3827 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...