Ethanesulphonic acid 95%
Supplier: Thermo Scientific
|
Danger
|
366040050
366040250
366041000
ACRO366040050EA
33.9
EUR
ACRO366040050
ACRO366040250
ACRO366041000
Ethanesulphonic acid 95%
Ethanesulphonic acid
Formula:
C₂H₆O₃S MW: 110,13 g/mol Boiling Pt: 136…140 °C (3hPa) Melting Pt: –17 °C Density: 1,343 g/cm³ Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007529 CAS Number: 594-45-6 EINECS: 209-843-0 UN: 2586 ADR: 8,III |
Specification Test Results
Appearance | Clear yellow to brown Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4325 to 1.4355 (20°C, 589 nm) |
Titration with NaOH | ≥94 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...