Methanesulphonic acid ≥98%
Supplier: Thermo Fisher Scientific
|
Danger
|
A13565.0E
A13565.22
A13565.36
A13565.0EEA
302
EUR
A13565.0E
A13565.22
A13565.36
Methanesulphonic acid ≥98%
Methanesulphonic acid
Purity: 98+%
CAS No.: 75-75-2
Molecular Formula: CH4O3S
Formula Weight: 96.10
Flash Point: 156°(312°F)
Refractive Index: 1.4300
Melting Point: 19-20°
Boiling Point: 166-168°/10mm
Beilstein Reference: 1446024
Merck Reference: 14,5954
UN No.: 2586
MDL No.: MFCD00007518
Formula:
CH₃SO₃H MW: 96.11 g/mol Boiling Pt: 166…168 °C (10 mmHg) Melting Pt: 19…20 °C Density: 1.481 Flash Pt: 156 °C (312 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007518 CAS Number: 75-75-2 EINECS: 200-898-6 UN: 2586 ADR: 8 III Merck Index: 13,05981 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...