n-Pentane 98%, Reagent Grade
Supplier: Honeywell Chemicals
|
Danger
|
158941-2.5L
158941-1L
158941-2.5LEA
148
EUR
158941-2.5L
158941-1L
n-Pentane 98%, Reagent Grade
n-Pentane
- ≥95% for general laboratory use.
Beilstein Number: 969132
RTECS Number: RZ9450000
UNSPSC Code: 12191502
Formula:
H₃C(CH₂)₃CH₃ Boiling Pt: 36-37 °C Melting Pt: -131 °C Density: 0,631 g/cm³ (15 °C) Flash Pt: <-35 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009498 CAS Number: 109-66-0 EINECS: 203-692-4 UN: 1265 ADR: 3,II REACH: 01-2119459286-30 Merck Index: 13,07193 |
|
Specification Test Results
Vapor Density | 2.48 (vs air) |
Assay (GC) | Min. 97.5 % |
Refractive index (n 20/D) | 1.356 - 1.360 |
IR spectrum | complying |
Refractive Index | n20/D 1.358 (lit.) |
Explosion Limit | ~8.3 % |
Autoignition Temperature | 500 °F |
Vapor Pressure | 8.43 psi ( 20 °C) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...