1,2-Epoxy-3-methylbutane ≥98%
Supplier: Thermo Scientific
|
Danger
|
L19291.03
L19291.06
L19291.03EA
70.4
EUR
L19291.03
L19291.06
1,2-Epoxy-3-methylbutane ≥98%
1,2-Epoxy-3-methylbutane
Purity: 98+%
CAS No.: 1438-14-8
Molecular Formula: C5H10O
Formula Weight: 86.13
Flash Point: -12°(10°F)
Refractive Index: 1.3915
Odour: Recognizable
Boiling Point: 72-75°
Beilstein Reference: 102418
UN No.: 1993
MDL No.: MFCD02683501
Formula:
C₅H₁₀O MW: 86.13 g/mol Boiling Pt: 72…75 °C Density: 0,815 Flash Pt: –12 °C (10 °F) Storage Temperature: Ambient |
MDL Number:
MFCD02683501 CAS Number: 1438-14-8 EINECS: 215-869-3 UN: 1993 ADR: 3,II |
Learn more
![](/m-ie.vwr.com/en_IE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...