Methyl pivalate 99%

Supplier: Thermo Scientific
Danger

Synonyms: Methyl trimethylacetate

127952500 127951000
ACRO127952500EA 61.5 EUR
ACRO127952500 ACRO127951000
Methyl pivalate 99%
Methyl pivalate
Formula: C₆H₁₂O₂
MW: 116,16 g/mol
Boiling Pt: 101 °C (760 mmHg)
Melting Pt: –70 °C
Density: 0,87 g/cm³
Flash Pt: <1 °C
Storage Temperature: Ambient
MDL Number: MFCD00008843
CAS Number: 598-98-1
EINECS: 209-959-1
UN: 3272
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3890 to 1.3910 (20°C, 589 nm)
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR