3,3,5-Trimethylcaproic acid ≥97%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
3,3,5-Trimethylhexanoic acid, 3,5,5-Trimethylhexanoicacid, 3,5,5-Trimethylhexanoic acid
L13360.AE
L13360.AP
L13360.AEEA
45.4
EUR
L13360.AE
L13360.AP
3,3,5-Trimethylcaproic acid ≥97%
3,3,5-Trimethylcaproic acid
Purity: 97%
CAS No.: 3302-10-1
Molecular Formula: C9H18O2
Formula Weight: 158.24
Flash Point: 125°(257°F)
Refractive Index: 1.4290
Odour: Unpleasant
Melting Point: -70°
Boiling Point: 230-232°
Beilstein Reference: 1749506
MDL No.: MFCD00020507
Formula:
C₉H₁₈O₂ MW: 158,24 g/mol Boiling Pt: 230…232 °C Melting Pt: –70 °C Density: 0,910 Flash Pt: 125 °C (257 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00020507 CAS Number: 3302-10-1 EINECS: 221-975-0 UN: 3265 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...