Bis(2-ethylhexyl)amine 99%
Supplier: Thermo Fisher Scientific
|
Danger
|
Synonyms:
Dioctylamine, Di(2-ethylhexyl)amine, Bis(2-ethylhexyl)amine
271960010
271962500
271960025
ACRO271960010EA
153
EUR
ACRO271960010
ACRO271962500
ACRO271960025
Bis(2-ethylhexyl)amine 99%
Bis(2-ethylhexyl) amine
Formula:
C₁₆H₃₅N MW: 241.46 g/mol Boiling Pt: 281.1 Melting Pt: –60 °C Density: 0.8 g/cm³ Flash Pt: 128 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009489 CAS Number: 106-20-7 EINECS: 203-372-4 UN: 2735 ADR: 8 II |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4415 to 1.4435 (20°C, 589 nm) |
GC | ≥98.5 % |
Titration with HClO4 | 98.5 to 101.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...