(4-Bromophenyl)acetic acid 99%

Supplier: Thermo Fisher Scientific
Warning

160520250 160521000
ACRO160520250EA 76.7 EUR
ACRO160520250 ACRO160521000
(4-Bromophenyl)acetic acid 99%
(4-Bromophenyl)acetic acid
Formula: (C₆H₄Br)CH₂COOH
Boiling Pt: 326 °C (1013 hPa)
Melting Pt: 112…114 °C
Storage Temperature: Ambient
MDL Number: MFCD00004342
CAS Number: 1878-68-8
EINECS: 217-523-7

Order Now

Specification Test Results

Appearance White to yellow to light beige Crystalline powder
Infrared spectrum Conforms
Melting point 115 to 118 °C
Titration with NaOH ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR