(3,4-Dichlorophenyl)acetic acid methyl ester ≥97%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
Methyl 3,4-Dichlorophenylacetate, 3,4-Dichlorophenylacetic acid methyll ester, Methyl 2-(3,4-dichlorophenyl)acetate, Methyl (3,4-dichlorophenyl)acetate
B21518.09
B21518.18
B21518.09EA
0
EUR
B21518.09
B21518.18
(3,4-Dichlorophenyl)acetic acid methyl ester ≥97%
(3,4-Dichlorophenyl)acetic acid methyl ester
Purity: 97+%
CAS No.: 6725-44-6
Molecular Formula: C9H8Cl2O2
Formula Weight: 219.07
Melting Point: 27-30°
MDL No.: MFCD00800680
Formula:
(C₆H₃Cl₂)CH₂COOCH₃ MW: 219,07 g/mol Melting Pt: 27…30 °C Storage Temperature: Ambient |
MDL Number:
MFCD00800680 CAS Number: 6725-44-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...