(2-Chlorophenyl)acetic acid 98.5+%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
2-Chlorophenylacetic acid
184030050
184030250
184031000
ACRO184030050EA
24.3
EUR
ACRO184030050
ACRO184030250
ACRO184031000
(2-Chlorophenyl)acetic acid 98.5+%
(2-Chlorophenyl)acetic acid
Formula:
(C₆H₄Cl)CH₂COOH MW: 170,6 g/mol Melting Pt: 94…97 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004317 CAS Number: 2444-36-2 EINECS: 219-482-0 |
Specification Test Results
Appearance | White to light yellow Powder |
Infrared spectrum | Conforms |
Melting point | 94 to 97 °C |
HPLC | ≥98.50 % |
Titration with TBAH | ≥98.50 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...