Methyl benzilate ≥98%
Supplier: Thermo Scientific
|
Warning
|
A15907.18
A15907.30
A15907.18EA
26.5
EUR
A15907.18
A15907.30
Methyl benzilate ≥98%
Methyl benzilate
Purity: 98%
CAS No.: 76-89-1
Molecular Formula: C15H14O3
Formula Weight: 242.28
Melting Point: 73-76°
Boiling Point: 187°/13mm
Merck Reference: 14,1080
MDL No.: MFCD00004446
Formula:
C₁₅H₁₄O₃ MW: 242,27 g/mol Boiling Pt: 187 °C (13 mmHg) Melting Pt: 73…76 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004446 CAS Number: 76-89-1 EINECS: 200-991-1 Merck Index: 14,1080 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...