2,4-Dimethylphenylacetic acid ≥98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
(2,4-Dimethylphenyl)acetic acid, 2,4-Xylylacetic acid
H50284.03
H50284.MD
H50284.03EA
265
EUR
H50284.03
H50284.MD
2,4-Dimethylphenylacetic acid ≥98%
2,4-Dimethylphenylacetic acid
Purity: 98%
CAS No.: 6331-04-0
Molecular Formula: C10H12O2
Formula Weight: 164.21
MDL No.: MFCD00014369
Formula:
(C₆H₃(CH₃)₂)CH₂COOH MW: 164,2 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD02664685 CAS Number: 6331-04-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...