3,5-Xylylacetic acid ≥98%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 2-(3,5-Dimethylphenyl)acetic acid, (3,5-Dimethylphenyl)acetic acid

L09707.03 L09707.06
L09707.03EA 55.2 EUR
L09707.03 L09707.06
3,5-Xylylacetic acid ≥98%
3,5-Xylylacetic acid

Purity: 98+% CAS No.: 42288-46-0 Molecular Formula: C10H12O2 Formula Weight: 164.21 Melting Point: 99-103° Beilstein Reference: 2083903

Formula: (C₆H₃(CH₃)₂)CH₂COOH
MW: 164.2 g/mol
Melting Pt: 99…103 °C
Storage Temperature: Ambient
MDL Number: MFCD00082776
CAS Number: 42288-46-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR