(4-Biphenyl)acetic acid 98%

Supplier: Thermo Scientific
Danger

Synonyms: 4-Biphenylacetic acid

188150250 188150100
ACRO188150250EA 164 EUR
ACRO188150250 ACRO188150100
(4-Biphenyl)acetic acid 98%
(4-Biphenyl)acetic acid
Formula: (C₆H₅C₆H₅)CH₂COOH
MW: 212,25 g/mol
Boiling Pt: 365 °C (1013 hPa)
Melting Pt: 156…160 °C
Storage Temperature: Ambient
MDL Number: MFCD00004351
CAS Number: 5728-52-9
EINECS: 227-233-2
UN: 2811
ADR: 6.1,III
Merck Index: 13,03979

Order Now

Specification Test Results

Appearance White to beige Powder
Infrared spectrum Conforms
Melting point 158 to 165 °C
Titration with TBAH ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR