(2-Benzyloxyphenyl)acetic acid ≥98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
2-(2-(Benzyloxy)phenyl)acetic acid
A15237.03
A15237.06
A15237.03EA
74.8
EUR
A15237.03
A15237.06
(2-Benzyloxyphenyl)acetic acid ≥98%
(2-Benzyloxyphenyl)acetic acid
Purity: 98+%
CAS No.: 22047-88-7
Molecular Formula: C15H14O3
Formula Weight: 242.28
Melting Point: 96-98°
Beilstein Reference: 2653977
Formula:
C₆H₅CH₂OC₆H₄CH₂COOH MW: 242,27 g/mol Melting Pt: 96…98 °C Storage Temperature: Ambient |
MDL Number:
MFCD00016822 CAS Number: 22047-88-7 UN: 3077 ADR: 9,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...