(2-Fluorophenyl)acetic acid ≥98%
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
2-(2-Fluorophenyl)acetic acid, 2-Fluorophenylacetic acid
A10596.09
A10596.18
A10596.09EA
55.6
EUR
A10596.09
A10596.18
(2-Fluorophenyl)acetic acid ≥98%
(2-Fluorophenyl)acetic acid
Purity: 98+%
CAS No.: 451-82-1
Molecular Formula: C8H7FO2
Formula Weight: 154.14
Odour: Odourless
Melting Point: 61-65°
Beilstein Reference: 775955
MDL No.: MFCD00004315
Formula:
(C₆H₄F)CH₂COOH Boiling Pt: 263 °C (1013 hPa) Melting Pt: 61…65 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004315 CAS Number: 451-82-1 EINECS: 207-196-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...