(4-Fluorophenyl)acetic acid 98%

Supplier: Thermo Scientific
Warning

Synonyms: 2-(4-Fluorophenyl)acetic acid

119570100 119570250 119571000
ACRO119570100EA 42.9 EUR
ACRO119570100 ACRO119570250 ACRO119571000
(4-Fluorophenyl)acetic acid 98%
(4-Fluorophenyl)acetic acid
Formula: (C₆H₄F)CH₂COOH
MW: 154,14 g/mol
Boiling Pt: 264 °C (1013 hPa)
Melting Pt: 85…86 °C
Storage Temperature: Ambient
MDL Number: MFCD00004343
CAS Number: 405-50-5
EINECS: 206-972-4
Merck Index: 13,04204

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 81 to 85 °C
HPLC ≥97.5 %
Stabilizer 0.1 % w/w hydroquinone

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR