(2-Iodophenyl)acetic acid ≥98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
2-(2-Iodophenyl)acetic acid
L15116.03
L15116.06
L15116.03EA
23.8
EUR
L15116.03
L15116.06
(2-Iodophenyl)acetic acid ≥98%
(2-Iodophenyl)acetic acid
Purity: 98+%
CAS No.: 18698-96-9
Molecular Formula: C8H7IO2
Formula Weight: 262.05
Melting Point: 115-117°
Beilstein Reference: 2086800
Formula:
(C₆H₄I)CH₂COOH MW: 262,05 g/mol Melting Pt: 115…117 °C Storage Temperature: Ambient |
MDL Number:
MFCD00046546 CAS Number: 18698-96-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...