Glyoxalbis(2-hydroxyanil) indicator for metal titration, Fluka™

Supplier: Honeywell Chemicals

Synonyms: 2,2'-Ethanediylidenedinitrilodiphenol, 2,2'-(Ethane-1,2-diylidenebis(azanylylidene))diphenol, Glyoxalbis(2-hydroxyanil), 1,2-Bis(2-hydroxyphenylimino)ethane

33255-25G 33255-6X25G
33255-25GEA 468 EUR
33255-25G 33255-6X25G
Glyoxalbis(2-hydroxyanil) indicator for metal titration, Fluka™
Glyoxalbis(2-hydroxyanil)
Formula: C₁₄H₁₂N₂O₂
MW: 240.26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00002183
CAS Number: 1149-16-2
EINECS: 214-560-0

Order Now

Specification Test Results

Appearance complying
Identity (IR) complying
Residue on ignition Max. 0.1 %
Suitable for metal definition complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR