trans-Dichloroethylene ≥98% stabilised
Supplier: Thermo Fisher Scientific
|
Danger
|
Synonyms:
(E)-1,2-Dichloroethene, trans-Dichloroethene, (E)-1,2-Dichloroethylene, trans-1,2-Dichloroethylene
Stabilisation:
Stabilised with hydroquinone monomethyl ether (4-methoxyphenol, MEHQ)
L13994.14
L13994.22
L13994.14EA
96.5
EUR
L13994.14
L13994.22
trans-Dichloroethylene ≥98% stabilised
trans-Dichloroethylene
Stabilized with 4-methoxyphenol
Purity: 98%
CAS No.: 156-60-5
Molecular Formula: C2H2Cl2
Formula Weight: 96.94
Flash Point: 2°(35°F)
Refractive Index: 1.4455
Melting Point: -50°
Boiling Point: 47-49°
Beilstein Reference: 1420761
Merck Reference: 14,92
UN No.: 1150
MDL No.: MFCD00062942
Formula:
C₂H₂Cl₂ Boiling Pt: 47…49 °C Melting Pt: –50 °C Density: 1,252 Flash Pt: 2 °C (35 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00062942 CAS Number: 156-60-5 EINECS: 205-860-2 UN: 1150 ADR: 3,II Merck Index: 14,92 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...